missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP170 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27325-1-AP-20UL
This item is not returnable.
View return policy
Description
The product of this gene is a component of the centrosome, a non-membraneous organelle that functions as the major microtubule-organizing center in animal cells. During interphase, the encoded protein localizes to the sub-distal appendages of mature centrioles, which are microtubule-based structures thought to help organize centrosomes. During mitosis, the protein associates with spindle microtubules near the centrosomes. The protein interacts with and is phosphorylated by polo-like kinase 1, and functions in maintaining microtubule organization and cell morphology. The human genome contains a putative transcribed pseudogene. Several alternatively spliced transcript variants of this gene have been found, but the full-length nature of some of these variants has not been determined.Specifications
| CEP170 | |
| Polyclonal | |
| Unconjugated | |
| CEP170 | |
| centrosomal protein 170kDa, Centrosomal protein of 170 kDa, CEP170, FAM68A, KAB, KARP 1 binding protein, KARP1 binding protein, KIAA0470 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9859 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunocytochemistry | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5SW79 | |
| CEP170 | |
| CEP170 Fusion Protein Ag26311 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction