missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CEP170 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583592
This item is not returnable.
View return policy
Description
Immunogen sequence: IPPLVHSKTP EGNNGRSGDP RPQAAEPPDH LTITRRRTWS RDEVMGDNLL LSSVFQFSKK IRQSIDKTAG KIRILFKDKD RNWDDIESKL RAESEVPIVK T Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 34%.
The product of this gene is a component of the centrosome, a non-membraneous organelle that functions as the major microtubule-organizing center in animal cells. During interphase, the encoded protein localizes to the sub-distal appendages of mature centrioles, which are microtubule-based structures thought to help organize centrosomes. During mitosis, the protein associates with spindle microtubules near the centrosomes. The protein interacts with and is phosphorylated by polo-like kinase 1, and functions in maintaining microtubule organization and cell morphology. The human genome contains a putative transcribed pseudogene. Several alternatively spliced transcript variants of this gene have been found, but the full-length nature of some of these variants has not been determined.
Specifications
| CEP170 | |
| Polyclonal | |
| Unconjugated | |
| CEP170 | |
| 4933426L22Rik; A330004A13Rik; AI195353; centrosomal protein 170; centrosomal protein 170kDa; centrosomal protein of 170 kDa; CEP170; FAM68A; KAB; KARP-1-binding protein; KARP1-binding protein; KIAA0470; XRCC5 binding protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9859 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5SW79 | |
| CEP170 | |
| Recombinant protein corresponding to Human CEP170. Recombinant protein control fragment (Product #RP-99492). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction