missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP170/CEP170L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | CEP170/CEP170L |
|---|---|
| Concentration | 0.4 mg/mL |
| Applications | Immunocytochemistry, Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16878734
|
Proteintech
18899-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16868734
|
Proteintech
18899-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This locus appears to be a transcribed pseudogene similar to centrosomal protein 170 kDa (CEP170). An approximately 50 kb region upstream of this locus also is homologous to CEP170, but is not transcribed.Specifications
| CEP170/CEP170L | |
| Immunocytochemistry, Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Rat, Human, Mouse | |
| Q6A065, Q96L14 | |
| 171457, 545389, 645455 | |
| CEP170/CEP170L Fusion Protein Ag13532 | |
| Primary | |
| -20°C | |
| CEP170P1 |
| 0.4 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| CEP170, CEP170L, FAM68B, KIAA0470, KIAA0470L | |
| Cep170, CEP170P1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title