missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP152 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21815-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the cytoskeletal BTB/kelch repeat family. The encoded protein plays a role in neurofilament architecture and is involved in mediating the ubiquitination and degradation of some proteins. Defects in this gene are a cause of giant axonal neuropathy.Specifications
| CEP152 | |
| Polyclonal | |
| Unconjugated | |
| CEP152 | |
| centrosomal protein 152kDa, Centrosomal protein of 152 kDa, CEP152, KIAA0912 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22995, 99100 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| A2AUM9, O94986 | |
| Cep152 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction