missing translation for 'onlineSavingsMsg'
Learn More

CEP-37440, MedChemExpress

Product Code. 16419407
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16419407 5 mg 5mg
16496956 100 mg 100mg
16426868 10 mg 10mg
16402937 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16419407 Supplier Medchem Express Supplier No. HY15841

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CEP-37440 is a novel potent and selective Dual FAK/ALK inhibitor with IC50 s of 2.3 nM (FAK) and 120 nM(ALK cellular IC50 in 75% human plasma).

CEP-37440 is a novel potent and selective Dual FAK/ALK inhibitor with IC50 s of 2.3 nM (FAK) and 120 nM(ALK cellular IC50 in 75% human plasma).

Specifications

Chemical Name or Material CEP-37440
CAS 1391712-60-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C30H38ClN7O3
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (172.38 mM; Need ultrasonic)
SMILES O=C(NC)C1=CC=CC=C1NC2=NC(NC3=CC=C4C(CCC[C@H](N5CCN(CCO)CC5)C4)=C3OC)=NC=C2Cl
Molecular Weight (g/mol) 580.12
Formula Weight 580.12
Percent Purity 99.97%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.