missing translation for 'onlineSavingsMsg'
Learn More

CEP-33779, MedChemExpress

Product Code. 16462567
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16462567 50 mg 50mg
16439057 5 mg 5mg
16426468 10 mg 10mg
16406746 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16462567 Supplier Medchem Express Supplier No. HY15343

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CEP-33779 is a novel, selective, and orally bioavailable inhibitor of JAK2 with an IC50 of 1.8±0.6 nM.

CEP-33779 is a novel, selective, and orally bioavailable inhibitor of JAK2 with an IC50 of 1.8±0.6 nM.

Specifications

Chemical Name or Material CEP-33779
CAS 1257704-57-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H26N6O2S
Quantity 50 mg
Solubility Information DMSO : 50 mg/mL (108.09 mM; Need ultrasonic)
SMILES O=S(C1=CC=C(C2=CC=CN3C2=NC(NC4=CC=CC(N5CCN(C)CC5)=C4)=N3)C=C1)(C)=O
Molecular Weight (g/mol) 462.57
Formula Weight 462.57
Percent Purity 99.36%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.