missing translation for 'onlineSavingsMsg'
Learn More

CEP-28122 mesylate salt, MedChemExpress

Product Code. 16499927
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499927 5 mg 5mg
16453477 50 mg 50mg
16407458 10 mg 10mg
16407426 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16499927 Supplier Medchem Express Supplier No. HY18030A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CEP-28122 mesylate salt, a diaminopyrimidine derivative, is a potent, selective, and orally bioavailable ALK inhibitor, with an IC50 value of 1.9 nM for recombinant ALK kinase activity. CEP-28122 has antitumor activity in experimental models of ALK-positive human cancers. CEP-28122 mesylate salt has good pharmacodynamic and pharmacokinetic activity.

CEP-28122 mesylate salt, a diaminopyrimidine derivative, is a potent, selective, and orally bioavailable ALK inhibitor, with an IC50 value of 1.9 nM for recombinant ALK kinase activity. CEP-28122 has antitumor activity in experimental models of ALK-positive human cancers. CEP-28122 mesylate salt has good pharmacodynamic and pharmacokinetic activity.

Specifications

Chemical Name or Material CEP-28122 mesylate salt
Color Off-White
Purity Grade Notes Research
Molecular Formula C29H39ClN6O6S
Quantity 5 mg
Solubility Information DMSO : ≥ 6.4 mg/mL (10.08 mM)
SMILES O=C([C@H]1[C@](C2)([H])C=C[C@]2([H])[C@H]1NC3=NC(NC4=CC=C5C(CC[C@@H](N6CCOCC6)CC5)=C4OC)=NC=C3Cl)N.O=S(C)(O)=O
Molecular Weight (g/mol) 635.17
Formula Weight 635.17
Percent Purity 99.85%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.