missing translation for 'onlineSavingsMsg'
Learn More

Centrophenoxine hydrochloride, 98%

Product Code. 15486259
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15486259 100 g 100g
15466259 5 g 5g
15476259 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15486259 Supplier Thermo Scientific Alfa Aesar Supplier No. J64196.22
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3685-84-5 | C12H17Cl2NO3 | 294.17 g/mol

Ester of dimethylethanolamine and 4-chlorophenoxyacetic acid; activates acetylcholinesterase activity in hippocampus of aged rats

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ester of dimethylethanolamine and 4-chlorophenoxyacetic acid; activates acetylcholinesterase activity in hippocampus of aged rats
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3685-84-5
Molecular Formula C12H17Cl2NO3
Molecular Weight (g/mol) 294.17
MDL Number MFCD00012533
InChI Key FIVHOHCAXWQPGC-UHFFFAOYSA-N
Synonym meclofenoxate hydrochloride, centrophenoxine hydrochloride, lucidril, cerutil, meclofenoxate hcl, atsefen, helfergin, marucotol, acefen, brenal
PubChem CID 19379
IUPAC Name hydrogen 2-(dimethylamino)ethyl 2-(4-chlorophenoxy)acetate chloride
SMILES [H+].[Cl-].CN(C)CCOC(=O)COC1=CC=C(Cl)C=C1

Specifications

Melting Point 135°C to 145°C
Color White
Quantity 100 g
Merck Index 14,5780
Formula Weight 294.17
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Centrophenoxine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.