Learn More
Labconco CentriZap Pro Strobe Light
Product Code. 30533440 Shop All Labconco Corporation Products
Change view
Click to view available options
Rotor System:
CentriVap Pro
Unit Size:
Each
Product Code. Rotor System Unit Size
30533440 CentriVap Pro Each
1 options
Product Code. 30533440 Supplier Labconco Supplier No. 8547200
  • ONLINE EXCLUSIVE PRICE
    £73.90
    £72.59 / Pack of 1
    Save £1.31
    1% Off
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The CentriZap™ Strobe Light enables users to view samples as if they were stationary while rotor is spinning, eliminating the need to pause a cycle.
  • For use with Labconco CentriVap Pro Vacuum Concentrators.
  • Includes mounting holster and hardware for attachment to the right side of the CentriVap Pro Vacuum Concentrator.
  • Shipping weight 2.0 lbs. (0.9 kg)
Specifications
Type Strobe Light
Rotor System CentriVap Pro