Learn More
Labconco CentriVap Cart
Product Code. 30533428 Shop All Labconco Corporation Products
Change view
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
Product Code. Material Unit Size
30533428 Stainless Steel Each
1 options
Product Code. 30533428 Supplier Labconco Supplier No. 8544000
  • ONLINE EXCLUSIVE PRICE
    £1130.00
    £1031.16 / Pack of 1
    Save £98.84
    8% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Powder-coated steel cart supports CentriVap Vacuum Concentrators, CentriVap Cold Traps, and vacuum pumps during use, storage, and transportation.
  • For use with Labconco CentriVap Pro and CentriVap Standard Vacuum Concentrator Systems.
  • Ergonomic design allows for easy access to all key components of the system.
  • Shipping weight 132.0 lbs. (59.9 kg)
Specifications
Material Stainless Steel
Type Trolley Cart
Rotor System CentriVap Pro