missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Centrin 3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38508-20ul
This item is not returnable.
View return policy
Description
Centrin 3 Polyclonal antibody specifically detects Centrin 3 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| Centrin 3 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:100 | |
| CDC31 yeast homolog, CEN3centrin, EF-hand protein, 3 (CDC31 homolog, yeast), centrin, EF-hand protein, 3, centrin, EF-hand protein, 3 (CDC31 yeast homolog), centrin-3, EF-hand superfamily member, MGC12502, MGC138245 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-167 of human Centrin 3 (NP_004356.2).,, Sequence:, MSLALRSELVVDKTKRKKRRELSEEQKQEIKDAFELFDTDKDEAIDYHELKVAMRALGFDVKKADVLKILKDYDREATGKITFEDFNEVVTDWILERDPHEEILKAFKLFDDDDSGKISLRNLRRVARELGENMSDEELRAMIEEFDKDGDGEINQEEFIAIMTGDI | |
| 20 μL | |
| Cell Cycle and Replication | |
| 1070 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction