missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Centrin 2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144878
This item is not returnable.
View return policy
Description
Immunogen sequence: MASNFKKANM ASSSQRKRMS PKPELTEE Antigen sequence identity: Mouse 86%; Rat 93%.
RNP2 a type I integral membrane protein found only in the rough endoplasmic reticulum. The encoded protein is part of an N-oligosaccharyl transferase complex that links high mannose oligosaccharides to asparagine residues found in the Asn-X-Ser/Thr consensus motif of nascent polypeptide chains. This protein is similar in sequence to the yeast oligosaccharyl transferase subunit SWP1.
Specifications
| Centrin 2 | |
| Polyclonal | |
| Unconjugated | |
| CETN2 | |
| 1110034A02Rik; AI326150; CALT; Caltractin; caltractin (20kD calcium-binding protein); Caltractin isoform 1; caltractin, 20 kDa calcium binding protein; CEN2; centrin 2; centrin, EF-hand protein, 2; Centrin2; centrin-2; Cetn2; EF hand protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1069 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P41208 | |
| CETN2 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 1-28. Recombinant protein control fragment (Product #RP-109947). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction