missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Centrin 2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £360.00
Specifications
| Antigen | Centrin 2 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Knockout Validated |
| Applications | Western Blot, Immunofluorescence, Gene Knock-Out |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18685572
|
Novus Biologicals
NBP2-92653-0.02ml |
0.02 mL |
£161.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18616152
|
Novus Biologicals
NBP2-92653-0.1ml |
0.1 mL |
£360.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Centrin 2 Polyclonal antibody specifically detects Centrin 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Gene Knock-OutSpecifications
| Centrin 2 | |
| Western Blot, Immunofluorescence, Gene Knock-Out | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, DNA Repair, Nucleotide Excision Repair | |
| PBS with 50% glycerol, pH7.3. | |
| 1069 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Knockout Validated | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Caltractin isoform 1, CEN2CALTcaltractin (20kD calcium-binding protein), centrin, EF-hand protein, 2, centrin-2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-172 of human CETN2 (NP_004335.1). MASNFKKANMASSSQRKRMSPKPELTEEQKQEIREAFDLFDADGTGTIDVKELKVAMRALGFEPKKEEIKKMISEIDKEGTGKMNFGDFLTVMTQKMSEKDTKEEILKAFKLFDDDETGKISFKNLKRVAKELGENLTDEELQEMIDEADRDGDGEVSEQEFLRIMKKTSLY | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title