missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Centrin 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£169.00 - £255.00
Specifications
| Antigen | Centrin 1 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18614146
|
Novus Biologicals
NBP2-99465-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18692726
|
Novus Biologicals
NBP2-99465-50ul |
50 μg |
£169.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Centrin 1 Polyclonal antibody specifically detects Centrin 1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Centrin 1 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| 0.2 um filtered solution in PBS | |
| 1068 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| calcium binding protein, caltractin, Caltractin isoform 2, CEN1centrin-1, centrin, EF-hand protein, 1, CETN, EF-hand protein | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Centrin 1. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title