missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Centrin 1 Antibody (CETN1/9880), Novus Biologicals™
Mouse Monoclonal Antibody
£195.00 - £368.00
Specifications
| Antigen | Centrin 1 |
|---|---|
| Clone | CETN1/9880 |
| Concentration | 0.2 mg/ml |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30303183
|
Novus Biologicals
NBP3-44584-20ug |
20 μg |
£195.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30300011
|
Novus Biologicals
NBP3-44584-100ug |
100 μg |
£368.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Centrin 1 Monoclonal antibody specifically detects Centrin 1 in Human samples. It is validated for ImmunohistochemistrySpecifications
| Centrin 1 | |
| 0.2 mg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| calcium binding protein, caltractin, Caltractin isoform 2, CEN1centrin-1, centrin, EF-hand protein, 1, CETN, EF-hand protein | |
| Recombinant fragment (around aa 1-172) of human Centrin 1 protein (Uniprot: Q12798). Exact sequence is proprietary. | |
| Primary | |
| Store at 4⪚C. |
| CETN1/9880 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Cell Cycle and Replication | |
| 10 mM PBS with 0.05% BSA | |
| 1068 | |
| IgG | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title