missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KARTELL Centrifuge Graduated Micro Test Tubes
Description
- Ideal for samples analysis.
- Depending on centrifuge being used, these tubes can withstand a centrifugal force of up to RCF 11.000.
- Flat cap with writing area.
- Art. 279 available in: neutral (00), blue (04), yellow (06), pink (13) and green (12).
- MATERIAL: PP
Specifications
Specifications
| Autoclavable | Autoclavable |
| Color | Assorted (Red, Yellow, Blue, Green, Orange) |
| Material | Polypropylene |
| No. per Pack | 1000 pc. |
| Packaging | 10 pack |
| Sterility | Non-sterile |
| Graduated | Yes |
| Closure Type | Flat Snap Cap |
| Fit for Purpose | Certified Quality |
| Bottom Shape | Conical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?