missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CENTG1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5140279

Product Code. 17804800

  • £387.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: AASTPVAGQA SNGGHTSDYS SSLPSSPNVG HRELRAEAAA VAGLSTPGSL HRAAKRRTSL FANRRGSDSE KRSLDS Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 99%.

The protein encoded by this gene belongs to the centaurin gamma-like family. It mediates anti-apoptotic effects of nerve growth factor by activating nuclear phosphoinositide 3-kinase. It is overexpressed in cancer cells, and promotes cancer cell invasion. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CENTG1
Polyclonal
Unconjugated
Agap2
AGAP2; AGAP-2; Arf GAP with GTP-binding protein-like, ANK repeat and PH domains 2; ArfGAP with GTPase domain, ankyrin repeat and PH domain 2; arf-GAP with GTPase, ANK repeat and PH domain-containing protein 2; centaurin gamma1; centaurin, gamma 1; centaurin-gamma-1; CENTG1; cnt-g1; FLJ16430; GGAP2; GTP-binding and GTPase activating protein 2; GTP-binding and GTPase-activating protein 2; KIAA0167; mKIAA0167; nuclear GTPase PIKE; phosphatidylinositol 3-kinase enhancer; phosphatidylinositol-3-kinase enhancer; phosphoinositide 3-kinase enhancer; Pike
Rabbit
Antigen affinity chromatography
RUO
116986
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q99490
Agap2
Recombinant protein corresponding to Human AGAP2. Recombinant protein control fragment (Product #RP-109502).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.