Learn More
Motic™ Phase Centering Telescope
Product Code. 11733205
Click to view available options
For Use With (Equipment):
AE30, AE31-EA Inverted Microscopes
Unit Size:
Each
Product Code. 11733205
Brand: Motic™ 1101000300836
Only null left
Add to basket
Add to basket
Centers phase rings for phase contrast applications. Motic™ Phase Centering Telescope replaces the use of an eyepiece to move the intermediate image plane to an observable point.
  • For use with Inverted AE30/AE31-EA Series Microscopes
  • Centers phase ring
  • Adjusts the the image plane to an observable point
  • For phase contrast applications
Order Info
dimProductType
Specifications
Type Phase Centering Telescope
For Use With (Application) Phase contrast
For Use With (Equipment) AE30, AE31-EA Inverted Microscopes
Quantity 1 Ea.