missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Centaurin beta 2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92701-0.1ml
This item is not returnable.
View return policy
Description
Centaurin beta 2 Polyclonal antibody specifically detects Centaurin beta 2 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Centaurin beta 2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| Arf GAP with coiled coil, ANK repeat and PH domains 2, ArfGAP with coiled-coil, ankyrin repeat and PH domains 2, centaurin, beta 2, Centaurin-beta-2, CENTB2centaurin-beta-2, CNT-B2, KIAA0041arf-GAP with coiled-coil, ANK repeat and PH domain-containing protein 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 50-160 of human ACAP2 (NP_036419.3). KAFCVANKQFMNGIRDLAQYSSNDAVVETSLTKFSDSLQEMINFHTILFDQTQRSIKAQLQNFVKEDLRKFKDAKKQFEKVSEEKENALVKNAQVQRNKQHEVEEATNILT | |
| 0.1 mL | |
| Signal Transduction | |
| 23527 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction