missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Centaurin alpha 2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92057-0.1ml
This item is not returnable.
View return policy
Description
Centaurin alpha 2 Polyclonal antibody specifically detects Centaurin alpha 2 in Human samples. It is validated for Western Blot
Specifications
| Centaurin alpha 2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| arf-GAP with dual PH domain-containing protein 2, ArfGAP with dual PH domains 2, CENTA2centaurin-alpha 2 protein, centaurin beta, centaurin, alpha 2, centaurin-alpha-2, cent-b, Cnt-a2, HSA272195 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-381 of human ADAP2 (NP_060874.1). MGDRERNKKRLLELLRAPDTGNAHCADCGAADPDWASYKLGIFICLNCCGVHRNFPDISRVKSVRLDFWDDSIVEFMIHNGNLRVKAKFEARVPAFYYIPQANDCLVLKEQWIRAKYERREFMADGETISLPGNREGFLWKRGRDNSQFLRRKFVLLAREGLLKYFTKEQGKSPKAVISIKDLNATFQTEKIGHPHGLQITYRRDGHTRNLFVYHESGKEIVDWFNALRAARLQYLKMAFPELPESELVPFLTRNYLKQGFMEKTGPKQKEPFKKRWFALDCHERRLLYYKNPLDAFEQGQVFLGNKEQGYEAYEDLPKGIRGNRWKAGLTIVTPERRFVLTCPSEKEQQEWLESLRGVLSSPLTPLNRLTASTESGRSSR | |
| 0.1 mL | |
| Signal Transduction | |
| 55803 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction