missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Centaurin alpha-1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569828
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%.
Centaurin alpha-1 (ADAP1, ArfGAP With Dual PH Domains 1) is a Protein Coding gene. Among its related pathways are B Cell Receptor Signaling Pathway (sino) and Arf6 signaling events. Gene Ontology (GO) annotations related to this gene include GTPase activator activity and inositol 1,3,4,5 tetrakisphosphate binding. An important paralog of this gene is ADAP2.
Specifications
| Centaurin alpha-1 | |
| Polyclonal | |
| Unconjugated | |
| ADAP1 | |
| 4930431P11Rik; ADAP1; Arf-GAP with dual PH domain-containing protein 1; ArfGAP with dual PH domains 1; CENTA1; centaurin, alpha 1; centaurin-alpha; centaurin-alpha-1; Cnt-a1; GC1L; GCS1L; inositol(1,3,4,5)tetrakisphosphate receptor; p42IP4; phosphatidylinositol-3,4,5-triphosphate binding protein; putative MAPK-activating protein PM25 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 231821 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| 0 | |
| ADAP1 | |
| synthetic peptide corresponding to a region of mouse Centaurin alpha-1. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction