missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Centaurin alpha 1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB300-907
This item is not returnable.
View return policy
Description
Centaurin alpha 1 Polyclonal specifically detects Centaurin alpha 1 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| Centaurin alpha 1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| arf-GAP with dual PH domain-containing protein 1, ArfGAP with dual PH domains 1, CENTA1centaurin-alpha-1, centaurin, alpha 1, centaurin-alpha, Centaurin-alpha-1, Cnt-a1, GCS1L, p42IP4, Putative MAPK-activating protein PM25 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Cross reacts with Human and Mouse. Expected from sequence similarity: Rat, Pig and Dog. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:32000 | |
| NP_006860. | |
| ADAP1 | |
| Peptide with sequence C-QEYAVEAHFKHKP corresponding to C-Terminus according to NP_006860.1. | |
| 0.1 mg | |
| Signal Transduction | |
| 11033 | |
| Human, Mouse, Rat, Pig | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction