missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CENPU Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100725
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CENPU.
The function of this protein remains unknown.
Specifications
| CENPU | |
| Polyclonal | |
| Unconjugated | |
| Cenpu | |
| 1700029A22Rik; CENP50; CENP-50; Cenpu; CENP-U; CENPU50; Centromere protein of 50 kDa; centromere protein U; ICEN24; Interphase centromere complex protein 24; KLIP1; KSHV latent nuclear antigen interacting protein 1; KSHV latent nuclear antigen-interacting protein 1; LOC617866 protein; MLF1 interacting protein; MLF1-interacting protein; MLF1IP; myeloid leukemia factor 1 interacting protein; PBIP1; Polo-box-interacting protein 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 71876, 79682 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q71F23, Q8C4M7 | |
| Cenpu | |
| A synthesized peptide derived from human CENPU(Accession Q71F23), corresponding to amino acid residues M1-G35. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction