missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CENPT Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-28744CL1
This item is not returnable.
View return policy
Description
CENPT Polyclonal antibody specifically detects CENPT in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| CENPT | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 80152 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| C16orf56, CENP-Tchromosome 16 open reading frame 56, centromere protein T, FLJ13111, FLJ43376, ICEN22, Interphase centromere complex protein 22 | |
| The immunogen recognized by this antibody maps to a region between residue 480 and 520 of human centromere protein T using the numbering given in entry NP_079358.3 (GeneID 80152). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction