missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CENPQ Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584229
This item is not returnable.
View return policy
Description
Immunogen sequence: VIMTILSNSI KEKEEIQYHL NFLKKRLLQQ CETLKVPPKK MEDLTNVSSL LNMERARDKA NEEGLALLQE EI Highest antigen sequence indentity to the following orthologs: Rat - 58%, Mouse - 68%.
CENPQ is a subunit of a CENPH-CENPI-associated centromeric complex that targets CENPA to centromeres and is required for proper kinetochore function and mitotic progression.
Specifications
| CENPQ | |
| Polyclonal | |
| Unconjugated | |
| CENPQ | |
| C6orf139; CENPQ; CENP-Q; Centromere protein Q | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55166 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7L2Z9 | |
| CENPQ | |
| Recombinant protein corresponding to Human CENPQ. Recombinant protein control fragment (Product #RP-106586). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction