missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CENPP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531186
This item is not returnable.
View return policy
Description
Recommended positive controls: K562, HL-60, mouse liver. Predicted reactivity: Pig (81%), Rhesus Monkey (98%), Bovine (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CENPP is a subunit of a CENPH.
Specifications
| CENPP | |
| Polyclonal | |
| Unconjugated | |
| CENPP | |
| 1700022C02Rik; 4921518G09Rik; Cenpp; CENP-P; Centromere protein P | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 401541, 66336 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q6IPU0, Q9CZ92 | |
| CENPP | |
| Recombinant protein encompassing a sequence within the center region of human CENPP. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction