missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CENPP Monoclonal Antibody (3G8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA523311
This item is not returnable.
View return policy
Description
Peptide Sequence: MDAELAEVRA LQAEIAALRR ACEDPPAPWE EKSRVQKSFQ AIHQFNLEGW KSSKDLKNQL GHLESELSFL STLTGINIRN HSKQTEDLTS TEMTEKSI.
CENPP is a subunit of a CENPH.
Specifications
| CENPP | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q6IPU0 | |
| CENPP | |
| CENPP (NP_001012267, 1 a.a. ~ 98 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 3G8 | |
| Unconjugated | |
| CENPP | |
| 1700022C02Rik; 4921518G09Rik; Cenpp; CENP-P; Centromere protein P | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 401541 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction