missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CENPO Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-76240CL1
This item is not returnable.
View return policy
Description
CENPO Polyclonal antibody specifically detects CENPO in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| CENPO | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 79172 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA, Western Blot | |
| CoraFluor 1 | |
| CENP-OMGC11266, centromere protein O, centromeric protein O, ICEN36, ICEN-36, Interphase centromere complex protein 36, MCM21R | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the center of human CENPO. The immunogen is located within amino acids 170 - 220 of CENPO. Amino Acid Squence: AYSGRKYQADRLQSDF | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction