missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CENPN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100721
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CENPN.
The centromere is a specialized chromatin domain, present throughout the cell cycle, that acts as a platform on which the transient assembly of the kinetochore occurs during mitosis. All active centromeres are characterized by the presence of long arrays of nucleosomes in which CENPA (MIM 117139) replaces histone H3 (see MIM 601128). CENPN is an additional factor required for centromere assembly.
Specifications
| CENPN | |
| Polyclonal | |
| Unconjugated | |
| Cenpn | |
| 2610510J17Rik; AI426416; ATM interactor; centromere protein N; AW545024; BM039; BM-309; C16orf60; Cenpn; CENP-N; Centromere protein N; Da2-27; Da2-4; FLJ13607; FLJ22660; ICEN32; Interphase centromere complex protein 32; RGD1310953; Unknown (protein for MGC:133728) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 55839, 72155 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q96H22, Q9CZW2 | |
| Cenpn | |
| A synthesized peptide derived from human CENPN(Accession Q96H22), corresponding to amino acid residues S277-P327. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction