missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CENPN, Mouse anti-Human, Clone: 4A5-1C11, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00055839-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: MDETVAEFIKRTILKIPMNELTTILKAWDFLSENQLQTVNFRQRKESVVQHLIHLCEEKRASISDAALLDIIYMQFHQHQKVWDVFQMSKGPGEDVDLFDMKQFKNSFKKILQRALKNVTVSFRETEENAVWIRIAWGTQYTKPNQYKPTYVVYYSQTPYAFTSSSMLRRNTPLLGQELEATGKIYLRQEEIILDITEMKKACNSpecifications
| CENPN | |
| Monoclonal | |
| Unconjugated | |
| centromere protein N | |
| BM039/C16orf60/CENP-N/FLJ13607/FLJ22660 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 4A5-1C11 | |
| In 1x PBS, pH 7.4 | |
| BC007334 | |
| CENPN | |
| BM039 (AAH07334, 1 a.a. ∼ 204 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 55839 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction