missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CENPM Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550777
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total CENPM protein.
The centromere is a specialized chromatin domain, present throughout the cell cycle, that acts as a platform on which the transient assembly of the kinetochore occurs during mitosis. All active centromeres are characterized by the presence of long arrays of nucleosomes in which CENPA replaces histone H3. CENPM is an additional factor required for centromere assembly.The centromere is a specialized chromatin domain, present throughout the cell cycle, that acts as a platform on which the transient assembly of the kinetochore occurs during mitosis. All active centromeres are characterized by the presence of long arrays of nucleosomes in which CENPA (MIM 117139) replaces histone H3 (see MIM 601128). CENPM is an additional factor required for centromere assembly (Foltz et al., 2006 [PubMed 16622419]).[supplied by OMIM].
Specifications
| CENPM | |
| Polyclonal | |
| Unconjugated | |
| CENPM | |
| 2610019I03Rik; AI853711; bK250D10.2; C22orf18; Cenpm; CENP-M; Centromere protein M; hypothetical protein LOC615953; ICEN39; Interphase centromere complex protein 39; MGC861; PANE1; proliferation associated nuclear element 1; proliferation-associated nuclear element protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79019 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1.6 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9NSP4 | |
| CENPM | |
| Full length fusion protein of Human CENPM. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction