missing translation for 'onlineSavingsMsg'
Learn More

CENPM Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£381.00

Specifications

Antigen CENPM
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18296882
View Documents
Novus Biologicals
NBP1-54361
100 μL
£381.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CENPM Polyclonal specifically detects CENPM in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

CENPM
Polyclonal
Rabbit
Q9NSP4
79019
Synthetic peptides corresponding to CENPM(centromere protein M) The peptide sequence was selected from the middle region of CENPM. Peptide sequence CDLEVEGFRATMAQRLVRVLQICAGHVPGVSALNLLSLLRSSEGPSLEDL.
Primary
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
bK250D10.2, C22orf18PANE1MGC861, CENP-Mchromosome 22 open reading frame 18, centromere protein M, ICEN39, Interphase centromere complex protein 39, Pane1, proliferation associated nuclear element 1, Proliferation-associated nuclear element protein 1
CENPM
IgG
20 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.