missing translation for 'onlineSavingsMsg'
Learn More

CENPK Antibody (4D11), Novus Biologicals™

Product Code. 18412579 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18412579 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18412579 Supplier Novus Biologicals Supplier No. H00064105M08

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CENPK Monoclonal antibody specifically detects CENPK in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen CENPK
Applications Western Blot, ELISA
Classification Monoclonal
Clone 4D11
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH05400
Gene Alias AF5alpha, CEMPK, CENP-KInterphase centromere complex protein 37, centromere protein K, FKSG14, ICEN37, leucine zipper protein FKSG14, P33, Protein AF-5alpha, SOLT, SoxLZ/Sox6-binding protein Solt
Host Species Mouse
Immunogen CENPK (AAH05400.1, 64 a.a. ∽ 155 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. VKCLTAELSQWQKKTPETIPLTEDVLITLGKEEFQKLRQDLEMVLSTKESKNEKLKEDLEREQRWLDEQQQIMESLNVLHSELKNKVETFSE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 64105
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.