Learn More
CENPJ Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-11517-100UL
This item is not returnable.
View return policy
Description
CENPJ, also named as CPAP, LAP and LIP1, belongs to the TCP10 family. It plays an important role in cell division and centrosome function by participating in centriole duplication. CENPJ inhibits microtubule nucleation from the centrosome.
Specifications
| CENPJ | |
| Polyclonal | |
| CoraLite 488 | |
| 100 μL | |
| 55835 | |
| Liquid |
| Immunofluorescence | |
| 1000 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| IgG |