missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CENPJ Polyclonal Antibody
CENPJ Polyclonal Antibody
Brand: Invitrogen PA566326
This item is not returnable.
View return policy
Description
CENPJ Polyclonal Antibody for ICC/IF
Specifications
| CENPJ | |
| Polyclonal | |
| Unconjugated | |
| CENPJ | |
| 4932437H03Rik; BM032; Cenpj; CENP-J; Centromere protein J; centrosomal P4.1-associated protein; CPAP; Gm81; LAG-3-associated protein; LAP; LIP1; LYST-interacting protein 1; LYST-interacting protein LIP1; LYST-interacting protein LIP7; MCPH6; microcephaly; Sas4; Sas-4; SASS4; SCKL4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55835 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9HC77 | |
| CENPJ | |
| Recombinant Human CENPJ. Recombinant protein control fragment (Product #RP-107007). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction