missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CENPH Antibody (OTI2G4), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-72163CL1
This item is not returnable.
View return policy
Description
CENPH Monoclonal antibody specifically detects CENPH in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF
Specifications
| CENPH | |
| Monoclonal | |
| CoraFluor 1 | |
| CENP-H, centromere protein H, ICEN35, Interphase centromere complex protein 35, kinetochore protein CENP-H, NNF1, PMF1 | |
| Full length human recombinant protein of human CENPH(NP_075060) produced in HEK293T cell. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 64946 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF | |
| OTI2G4 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction