missing translation for 'onlineSavingsMsg'
Learn More

CENPF Antibody (7F11.2), Novus Biologicals™

Product Code. 18358834 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18358834 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18358834 Supplier Novus Biologicals Supplier No. NBP313730

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CENPF Monoclonal antibody specifically detects CENPF in Human samples. It is validated for Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen CENPF
Applications Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 7F11.2
Conjugate Unconjugated
Formulation PBS, pH 7.2
Gene Alias AH antigen, cell-cycle-dependent 350K nuclear protein, CENF, CENP-F, CENP-F kinetochore protein, centromere protein F, centromere protein F (350/400kD, mitosin), centromere protein F, 350/400ka (mitosin), centromere protein F, 350/400kDa (mitosin), hcp-1, Kinetochore protein CENPF, mitosin, PRO1779
Host Species Mouse
Immunogen Protein fragment expressed in E. coli corresponding to amino acids 1759-2093.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 1063
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.