missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CENPE Antibody (CA8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43561
This item is not returnable.
View return policy
Description
CENPE Monoclonal antibody specifically detects CENPE in Cynomolgus Monkey,Human samples. It is validated for Flow Cytometry,ELISA
Specifications
| CENPE | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry, ELISA | |
| CENP-E, Centromere autoantigen E (312kD), Centromere protein E, centromere protein E (312kD), centromere protein E, 312kDa, centromere-associated protein E, KIF10, kinesin family member 10, Kinesin-related protein CENPE | |
| The original mouse parental antibody 118G03 was generated by immunizing SJL mice with recombinant human CENPE/TNFRSF17-Fc chimera (R&D 193-Fc) by using the RIMMS method (Rapid immunization multiple sites). A chimeric version of this antibody was generated and it is called CA8. (Uniprot# Q02224) | |
| 0.2 mg | |
| Cell Cycle and Replication | |
| 1062 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Flow Cytometry, ELISA | |
| CA8 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Cynomolgus Monkey, Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction