missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CENPC Recombinant Rabbit Monoclonal Antibody (23GB3760), Invitrogen™
GREENER_CHOICE

Product Code. 30087866
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30087866 100 μL 100µL
30089853 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30087866 Supplier Invitrogen™ Supplier No. MA552574

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Centromere protein C 1 is a centromere autoantigen and a component of the inner kinetochore plate. The protein is required for maintaining proper kinetochore size and a timely transition to anaphase. A putative pseudogene exists on chromosome 12.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CENPC
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3760
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CENPC
Gene Accession No. P49452, Q03188
Gene Alias CENPC; CENP-C; CENP-C 1; Cenpc1; Centromere autoantigen C; centromere autoantigen C1; centromere protein C; CENTROMERE PROTEIN C (CENP-C) (CENTROMERE AUTOANTIGEN C); centromere protein C 1; centromere protein C1; hcp-4; ICEN7; Interphase centromere complex protein 7; MIF2
Gene Symbols CENPC, Cenpc1
Host Species Rabbit
Immunogen A synthetic peptide of human CENPC (400-530AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1060, 12617, 305270
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.