Learn More
Invitrogen™ CENPC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109511
Description
Immunogen sequence: MAASGLDHLK NGYRRRFCRP SRARDINTEQ GQNVLEILQD CFEEKSLAND FSTNSTKSVP NSTRKIKDTC IQSPSKECQK SHPKSVPVSS KKKEASLQFV VEPSEATNRS VQAHEVHQKI LATDVSSKNT PDSKKISSRN INDHHSEADE EFYLSVGSPS VLLDAKTSVS QNVIPSSAQK RETYTFENSV NMLPSSTEVS VKTKKRLNFD DKVMLKKIEI DNKVSDEEDK TSEGQERKPS GSSQNRIRDS EYEIQRQAKK SFSTLFLETV KRKSESSPIV RHAATAPPHS CPPDDTKLIE.
Centromere protein C 1 is a centromere autoantigen and a component of the inner kinetochore plate. The protein is required for maintaining proper kinetochore size and a timely transition to anaphase. A putative pseudogene exists on chromosome 12.
Specifications
| CENPC | |
| Polyclonal | |
| Unconjugated | |
| CENPC | |
| CENPC; CENP-C; CENP-C 1; Cenpc1; Centromere autoantigen C; centromere autoantigen C1; centromere protein C; CENTROMERE PROTEIN C (CENP-C) (CENTROMERE AUTOANTIGEN C); centromere protein C 1; centromere protein C1; hcp-4; ICEN7; Interphase centromere complex protein 7; MIF2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1060, 12617, 305270 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.49 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P49452, Q03188 | |
| CENPC, Cenpc1 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.