missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CENPC Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566154
This item is not returnable.
View return policy
Description
Immunogen sequence: MSGKNDVDDEE VHGSSDDSKQ SKVIPKNRIH HKLVLPSNTP NVRRTKRTRL KPLEYWRGER IDYQGRPSGG FVISGVLSPD TISSKRK Highest antigen sequence identity to the following orthologs - mouse 48%, rat 53%.
Centromere protein C 1 is a centromere autoantigen and a component of the inner kinetochore plate. The protein is required for maintaining proper kinetochore size and a timely transition to anaphase. A putative pseudogene exists on chromosome 12.
Specifications
| CENPC | |
| Polyclonal | |
| Unconjugated | |
| CENPC | |
| CENPC; CENP-C; CENP-C 1; Cenpc1; Centromere autoantigen C; centromere autoantigen C1; centromere protein C; CENTROMERE PROTEIN C (CENP-C) (CENTROMERE AUTOANTIGEN C); centromere protein C 1; centromere protein C1; hcp-4; ICEN7; Interphase centromere complex protein 7; MIF2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1060 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q03188 | |
| CENPC | |
| Recombinant Human CENPC. Recombinant protein control fragment (Product #RP-106838). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction