missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CENPB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543753
This item is not returnable.
View return policy
Description
Peptide sequence: GGEDSDSDSE EEDDEEEDDE DEDDDDDEED GDEVPVPSFG EAMAYFAMVK Sequence homology: Cow: 83%; Horse: 86%; Human: 100%; Mouse: 93%; Pig: 86%; Rat: 100%; Sheep: 86%; Yeast: 100%; Zebrafish: 75%.
CENPB is a highly conserved protein that facilitates centromere formation. It is a DNA-binding protein that is derived from transposases of the pogo DNA transposon family. It contains a helix-loop-helix DNA binding motif at the N-terminus, and a dimerization domain at the C-terminus. It is also considered a major centromere autoantigen recognized by sera from patients with anti-centromere antibodies. This gene product is a highly conserved protein that facilitates centromere formation. It is a DNA-binding protein that is derived from transposases of the pogo DNA transposon family. It contains a helix-loop-helix DNA binding motif at the N-terminus, and a dimerization domain at the C-terminus. The DNA binding domain recognizes and binds a 17-bp sequence (CENP-B box) in the centromeric alpha satellite DNA. This protein is proposed to play an important role in the assembly of specific centromere structures in interphase nuclei and on mitotic chromosomes and is considered a major centromere autoantigen recognized by sera from patients with anti-centromere antibodies.
Specifications
| CENPB | |
| Polyclonal | |
| Unconjugated | |
| CENPB | |
| CENPB; CENP-B; CENP-B protein; centromere autoantigen B; centromere protein B; centromere protein B, 80kDa; major centromere autoantigen B | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1059 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P07199 | |
| CENPB | |
| Synthetic peptide directed towards the C-terminal of human CENPB (aa 506-555). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction