missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CENPB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565869
This item is not returnable.
View return policy
Description
Immunogen sequence: TFREKSRIIQE VEENPDLRKG EIARRFNIPP STLSTILKNK RAILASERKY GVASTCRKTN KLSPYDKLEG LLIAWFQQIR AAGLPVKGII L Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
CENPB is a highly conserved protein that facilitates centromere formation. It is a DNA-binding protein that is derived from transposases of the pogo DNA transposon family. It contains a helix-loop-helix DNA binding motif at the N-terminus, and a dimerization domain at the C-terminus. It is also considered a major centromere autoantigen recognized by sera from patients with anti-centromere antibodies. This gene product is a highly conserved protein that facilitates centromere formation. It is a DNA-binding protein that is derived from transposases of the pogo DNA transposon family. It contains a helix-loop-helix DNA binding motif at the N-terminus, and a dimerization domain at the C-terminus. The DNA binding domain recognizes and binds a 17-bp sequence (CENP-B box) in the centromeric alpha satellite DNA. This protein is proposed to play an important role in the assembly of specific centromere structures in interphase nuclei and on mitotic chromosomes and is considered a major centromere autoantigen recognized by sera from patients with anti-centromere antibodies.
Specifications
| CENPB | |
| Polyclonal | |
| Unconjugated | |
| CENPB | |
| CENPB; CENP-B; CENP-B protein; centromere autoantigen B; centromere protein B; centromere protein B, 80kDa; major centromere autoantigen B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1059 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Western Blot, Immunocytochemistry | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P07199 | |
| CENPB | |
| Recombinant Human CENPB. Recombinant protein control fragment (Product #RP-106539). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction