missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CENPB Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35700-100ul
This item is not returnable.
View return policy
Description
CENPB Polyclonal antibody specifically detects CENPB in Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| CENPB | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| CENP-B, centromere autoantigen B, Centromere protein B, centromere protein B (80kD), centromere protein B, 80kDa, major centromere autoantigen B | |
| A synthetic peptide corresponding to a sequence within amino acids 500 to the C-terminus of human CENPB (NP_001801.1).,, Sequence:, TLHFLEGGEDSDSDSEEEDDEEEDDEDEDDDDDEEDGDEVPVPSFGEAMAYFAMVKRYLTSFPIDDRVQSHILHLEHDLVHVTRKNHARQAGVRGLGHQS | |
| 100 μL | |
| Cell Cycle and Replication, Chromatin Research, DNA Repair | |
| 1059 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction