missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CENPA Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26754-1-AP-20UL
This item is not returnable.
View return policy
Description
Centromeres are the differentiated chromosomal domains that specify the mitotic behavior of chromosomes. CENPA encodes a centromere protein which contains a histone H3 related histone fold domain that is required for targeting to the centromere. CENPA is proposed to be a component of a modified nucleosome or nucleosome-like structure in which it replaces 1 or both copies of conventional histone H3 in the (H3-H4)2 tetrameric core of the nucleosome particle. Alternative splicing results in multiple transcript variants encoding distinct isoforms.Specifications
| CENPA | |
| Polyclonal | |
| Unconjugated | |
| CENPA | |
| CENP A, CENPA, Centromere autoantigen A, centromere protein A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1058 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P49450 | |
| CENPA | |
| CENPA Fusion Protein Ag25025 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction