missing translation for 'onlineSavingsMsg'
Learn More

CENPA Antibody (1D4-1A3), Novus Biologicals™

Product Code. 18390148 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18390148 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18390148 Supplier Novus Biologicals Supplier No. H00001058M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CENPA Monoclonal antibody specifically detects CENPA in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen CENPA
Applications ELISA
Classification Monoclonal
Clone 1D4-1A3
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH00881
Gene Alias Centromere autoantigen A, centromere protein A (17kD), centromere protein ACENP-Acentromere protein A, 17kDa, histone H3-like centromeric protein A
Host Species Mouse
Immunogen CENPA (AAH00881, 1 a.a. ~ 114 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MGPRRRSRKPEAPRRRSPSPTPTPGPSRRGPSLGASSHQHSRRRQGWLKEIRKLQKSTHLLIRKLPFSRLAAEAFLVHLFEDAYLLTLHAGRVTLFPKDVQLARRIRGLEEGLG
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 1058
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.