missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEND1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £514.00
Specifications
| Antigen | CEND1 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18476071
|
Novus Biologicals
NBP2-14469-25ul |
25ul |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18010654
|
Novus Biologicals
NBP2-14469 |
0.1 mL |
£514.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CEND1 Polyclonal specifically detects CEND1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CEND1 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| BM88 antigen, cell cycle exit and neuronal differentiation 1, cell cycle exit and neuronal differentiation protein 1, FLJ90066 | |
| CEND1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 51286 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: TTAPAKKTSAKADPALLNNHSNLKPAPTVPSSPDATPEPKGPGDGAEEDEAASGGPGGRGPWSCEN | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title