missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CEND Recombinant Rabbit Monoclonal Antibody (1D3Z1)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA538146
This item is not returnable.
View return policy
Description
Positive Samples: SH-SY5Y, U-87MG, Mouse brain, Rat brain Immunogen sequence: ALLNNHSNLK PAPTVPSSPD ATPEPKGPGD GAEEDEAASG GPGGRGPWSC ENFNPLLVAG GVAVAAIALI LGVAFLVRKK.
The protein encoded by this gene is a neuron-specific protein. The similar protein in pig enhances neuroblastoma cell differentiation in vitro and may be involved in neuronal differentiation in vivo. Multiple pseudogenes have been reported for this gene.
Specifications
| CEND | |
| Recombinant Monoclonal | |
| 0.54 mg/mL | |
| PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3 | |
| Q5FVI4, Q8N111, Q9JKC6 | |
| CEND1 | |
| Synthetic peptide | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry | |
| 1D3Z1 | |
| Unconjugated | |
| CEND1 | |
| 1500001H12Rik; AI415214; BM88; BM88 antigen; C38; cell cycle exit and neuronal differentiation 1; cell cycle exit and neuronal differentiation protein 1; CEND1; RGD1309401 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 361675, 51286, 57754 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction