missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CEND Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569458
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Dog: 86%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%.
The protein encoded by this gene is a neuron-specific protein. The similar protein in pig enhances neuroblastoma cell differentiation in vitro and may be involved in neuronal differentiation in vivo. Multiple pseudogenes have been reported for this gene.
Specifications
| CEND | |
| Polyclonal | |
| Unconjugated | |
| CEND1 | |
| 1500001H12Rik; AI415214; BM88; BM88 antigen; C38; cell cycle exit and neuronal differentiation 1; cell cycle exit and neuronal differentiation protein 1; CEND1; RGD1309401 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 51286 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N111 | |
| CEND1 | |
| synthetic peptide directed towards the N terminal region of human CEND. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction