missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CELSR3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533425
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon (100%) Monkey (89%) Marmoset (84%).
CELSR3 is a member of the flamingo subfamily, part of the cadherin superfamily. The flamingo subfamily consists of nonclassic-type cadherins; a subpopulation that does not interact with catenins. The flamingo cadherins are located at the plasma membrane and have nine cadherin domains, seven epidermal growth factor-like repeats and two laminin A G-type repeats in their ectodomain. They also have seven transmembrane domains, a characteristic unique to this subfamily. It is postulated that these proteins are receptors involved in contact-mediated communication, with cadherin domains acting as homophilic binding regions and the EGF-like domains involved in cell adhesion and receptor-ligand interactions. The specific function of this particular member has not been determined.
Specifications
| CELSR3 | |
| Polyclonal | |
| Unconjugated | |
| CELSR3 | |
| Adgrc3; adhesion G protein-coupled receptor C3; anchor protein; cadherin EGF LAG seven-pass G-type receptor 3; Cadherin family member 11; cadherin, EGF LAG seven-pass G-type receptor 3; cadherin, EGF LAG seven-pass G-type receptor 3 (flamingo homolog, Drosophila); CDHF11; Celsr3; EGFL1; EGF-like protein 1; EGF-like-domain, multiple 1; Epidermal growth factor-like protein 1; flamingo; flamingo homolog 1; FMI1; hFmi1; KIAA0812; Megf2; Megf3; mKIAA0812; multiple EGF-like domains protein 2; multiple epidermal growth factor-like domains 2; multiple epidermal growth factor-like domains protein 2; RESDA1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1951 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q9NYQ7 | |
| CELSR3 | |
| Synthetic 19 amino acid peptide from N-terminal extracellular domain of human CELSR3. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction